Learn More
Trajan Scientific CTC Analytics PAL System Needles For DLW Option, 22g, Pt. 3, 51mm, Without Locking Nut, 3 Pack
Catalog No. NC0612036
Encompass_Preferred
CTC Analytics PAL System Needles, For DLW Option, 22g, Pt. 3, 51mm, 3 Pack; Hamilton Part Number: 209221; For Syringe Part Numbers: 100.SyringeCDLW100R, LMK.2620735-BL, LMK.2620835-BL, LMK.2620935-BL, LMK.2620740, *Does not have nut included
Catalog No. NC0612036 Supplier Trajan Scientific Supplier No. PAL.DLWNDLX
CTC Analytics PAL System Needles, For DLW Option, 22g, Pt. 3, 51mm, 3 Pack; Hamilton Part Number: 209221; For Syringe Part Numbers: 100.SyringeCDLW100R, LMK.2620735-BL, LMK.2620835-BL, LMK.2620935-BL, LMK.2620740, *Does not have nut included