Learn More
Aviva Systems Biology Corp DLX2 Antibody - C-terminal region (ARP32212_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32212_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against DLX2. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human DLX2. Alternate names: TES-1, TES1ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rat
Catalog No. 50-014-54487