Learn More
Aviva Systems Biology Corp DLX3 Antibody - C-terminal region (ARP38584_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38584_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against DLX3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human DLX3. Alternate names: AI4, TDOApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Yeast
Catalog No. 50-014-56712