Learn More
Aviva Systems Biology Corp DLX3 Antibody - N-terminal region (ARP31605_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31605_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DLX3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human DLX3. Alternate names: TDO, AI4ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep
Catalog No. 50-014-54279