Learn More
Aviva Systems Biology Corp DLX4 Antibody - middle region (ARP34507_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34507_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DLX4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human DLX4. Alternate names: BP1, DLX7, DLX8, DLX9ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-55427