Learn More
Aviva Systems Biology Corp Dlx4 Antibody - middle region (ARP36838_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36838_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Dlx4. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: Dlx-4, Dlx7, MGC129167ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-284-520