Learn More
Aviva Systems Biology Corp DLX4 Antibody - N-terminal region (ARP39812_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39812_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DLX4. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human DLX4. Alternate names: BP1, DLX7, DLX8, DLX9ApplicationWB. Species reactivity: Cow, Human, Rat
Catalog No. 50-284-832