Learn More
Aviva Systems Biology Corp DLX5 Antibody - middle region (ARP32867_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32867_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DLX5. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human DLX5. Alternate names: SHFM1DApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-54800