Learn More
Aviva Systems Biology Corp Dlx6 Antibody - C-terminal region (ARP35763_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35763_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Dlx6. It was validated on Western Blot. . Alternate names: RGD1561539ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-284-473