Learn More
Cospheric LLC Density Marker Beads, Blue, 1.08g/cc, 20% concentration, 2.5 mL
SDP
Catalog No. NC1927114
Encompass
Density marker beads are small colored microspheres of known mass density that are used for calibrating density gradients and determining density in gradient columns.
Catalog No. NC1927114 Supplier Cospheric LLC Supplier No. DMBRBLU1.08 2.5ML
Density marker beads are small colored microspheres of known mass density that are used for calibrating density gradients and determining density in gradient columns.