Learn More
Supply Solutions SCILOGEX DM0412 LCD Digital Clinical Centrifuge, with 10200316 Rotor, & 19200317 Rotor Adapters & 19200318 Adapter Plugs: capacity 8 x 15ml / 12 x 10ml/7ml/5ml vacutainer, 110-220V, 50/60Hz, US Plug
Catalog No. NC0955229
Encompass_Preferred
The DM0412 Clinical Centrifuge is ideal for the separation of serum, plasma, urea, blood samples and other routine applications in hospital and research laboratories. The DM0412 conforms to international safety standards and regulations. - SCILOGEX clinical centrifuges have passed the explosion-proof test and are certified with CE, cTUVus and FCC. - MCA test according to IEC/EN61010-2-20 including explosion-proof and bio-safe tests - Passed EN61010-2-101:2002 requirements for in vitro diagnostic (IVD) medical equipment DM4124 Hematocrit Centrifuge Features: - High accuracy of speed, excellent performance - CPU controls all operating parameters including speed and time - RPM or G-force can be set and displayed - Operation can be timed from 30 seconds to 99 minutes or continuous running - Timer commences once the set speed is reached, so the separation time is more accurate - Gentle braking at low speeds with efficient separation - Paramete
Catalog No. NC0955229 Supplier Supply Solutions Supplier No. SCILOGEX 91302341
The DM0412 Clinical Centrifuge is ideal for the separation of serum, plasma, urea, blood samples and other routine applications in hospital and research laboratories. The DM0412 conforms to international safety standards and regulations. - SCILOGEX clinical centrifuges have passed the explosion-proof test and are certified with CE, cTUVus and FCC. - MCA test according to IEC/EN61010-2-20 including explosion-proof and bio-safe tests - Passed EN61010-2-101:2002 requirements for in vitro diagnostic (IVD) medical equipment DM4124 Hematocrit Centrifuge Features: - High accuracy of speed, excellent performance - CPU controls all operating parameters including speed and time - RPM or G-force can be set and displayed - Operation can be timed from 30 seconds to 99 minutes or continuous running - Timer commences once the set speed is reached, so the separation time is more accurate - Gentle braking at low speeds with efficient separation - Paramete