Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dm1-PEG4-DBCO | 1198.79 | C63H80ClN5O16 | 1 MG
SDP

Catalog No. 5000203904
Encompass_Preferred

DM1-PEG4-DBCO is a drug-linker conjugate that links the microtubule inhibitor DM1 to a DBCO-PEG4-Ahx linker for use in antibody-drug conjugate synthesis and bioorthogonal conjugation via strain-promoted alkyne-azide cycloaddition (SPAAC).

  • Drug-linker conjugate for antibody-drug conjugate synthesis.
  • Contains a DBCO group for strain-promoted alkyne-azide cycloaddition (SPAAC) click chemistry.
  • Molecular weight 1198.79 g/mol.
  • Chemical formula C63H80ClN5O16.
  • Available in small vials for research use, including 1 mg and 5 mg sizes.

Catalog No. 50-002-03904 Supplier Medchemexpress LLC Supplier No. HY1362611MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DM1-PEG4-DBCO is a drug-linker conjugate that links the microtubule inhibitor DM1 to a DBCO-PEG4-Ahx linker for use in antibody-drug conjugate synthesis and bioorthogonal conjugation via strain-promoted alkyne-azide cycloaddition (SPAAC).

  • Drug-linker conjugate for antibody-drug conjugate synthesis.
  • Contains a DBCO group for strain-promoted alkyne-azide cycloaddition (SPAAC) click chemistry.
  • Molecular weight 1198.79 g/mol.
  • Chemical formula C63H80ClN5O16.
  • Available in small vials for research use, including 1 mg and 5 mg sizes.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.