Learn More
Medchemexpress LLC DM1-PEG4-DBCO | 1198.79 g·mol⁻¹ | C63H80ClN5O16 | 10 MG
SDP
Catalog No. 5000202703
Encompass_Preferred
DM1-PEG4-DBCO is an agent-linker conjugate that combines the potent microtubule inhibitor DM1 with a DBCO-PEG4-Ahx linker to enable antibody-drug conjugate (ADC) synthesis via strain-promoted alkyne-azide cycloaddition (SPAAC). It is supplied as a solid reagent for research use only.
  • Agent-linker conjugate combining DM1 with a DBCO-PEG4-Ahx linker.
  • Designed for antibody-drug conjugate synthesis and bioconjugation.
  • Contains DBCO for strain-promoted alkyne-azide cycloaddition (SPAAC).
  • Enables conjugation to azide-containing biomolecules.
  • Supplied as a solid reagent; store protected from light at -20°C.
Catalog No. 50-002-02703 Supplier Medchemexpress LLC Supplier No. HY13626110MG
DM1-PEG4-DBCO is an agent-linker conjugate that combines the potent microtubule inhibitor DM1 with a DBCO-PEG4-Ahx linker to enable antibody-drug conjugate (ADC) synthesis via strain-promoted alkyne-azide cycloaddition (SPAAC). It is supplied as a solid reagent for research use only.
  • Agent-linker conjugate combining DM1 with a DBCO-PEG4-Ahx linker.
  • Designed for antibody-drug conjugate synthesis and bioconjugation.
  • Contains DBCO for strain-promoted alkyne-azide cycloaddition (SPAAC).
  • Enables conjugation to azide-containing biomolecules.
  • Supplied as a solid reagent; store protected from light at -20°C.