Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dm4-smcc | 1228105-52-9 | 96.9% | 1114.69 | C54H72ClN5O16S | 25 MG
SDP

Catalog No. 5000203965
Encompass_Preferred

DM4-SMCC is a drug-linker conjugate used as a cytotoxic payload in antibody-drug conjugate (ADC) research. It couples the maytansinoid DM4 to a non-cleavable SMCC linker, producing a stable antitumor agent supplied as a solid for laboratory use.

  • Drug-linker conjugate suitable for ADC research.
  • Contains a DM4 maytansinoid payload linked via a non-cleavable SMCC linker.
  • High purity, typically 96.9%.
  • Solid form, white to off-white powder; store frozen for long term stability.
  • Molecular weight about 1114.69 g/mol and formula C54H72ClN5O16S.
  • For research use only; not for human or veterinary use.

Catalog No. 50-002-03965 Supplier Medchemexpress LLC Supplier No. HY12649425MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DM4-SMCC is a drug-linker conjugate used as a cytotoxic payload in antibody-drug conjugate (ADC) research. It couples the maytansinoid DM4 to a non-cleavable SMCC linker, producing a stable antitumor agent supplied as a solid for laboratory use.

  • Drug-linker conjugate suitable for ADC research.
  • Contains a DM4 maytansinoid payload linked via a non-cleavable SMCC linker.
  • High purity, typically 96.9%.
  • Solid form, white to off-white powder; store frozen for long term stability.
  • Molecular weight about 1114.69 g/mol and formula C54H72ClN5O16S.
  • For research use only; not for human or veterinary use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.