Learn More
Medchemexpress LLC Dm4-sme | 796073-68-2 | 98.0% | 826.46 g/mol | C39H56ClN3O10S2 | 10 MG
SDP
Catalog No. 5000202813
Encompass_Preferred
DM4-SMe is a cytotoxic maytansinoid metabolite and microtubule/tubulin inhibitor used as an antibody-drug conjugate (ADC) payload in laboratory research. Supplied as a powder in small mass quantities, it is intended for in vitro and conjugation studies.
  • Cytotoxic maytansinoid metabolite and tubulin inhibitor.
  • Suitable as an ADC payload or for conjugation studies.
  • Supplied as a powder in small mass quantities for research use.
  • CAS number 796073-68-2 for unambiguous identification.
  • Reported molecular weight 826.46 g/mol and formula C39H56ClN3O10S2.
Catalog No. 50-002-02813 Supplier Medchemexpress LLC Supplier No. HY13008210MG
DM4-SMe is a cytotoxic maytansinoid metabolite and microtubule/tubulin inhibitor used as an antibody-drug conjugate (ADC) payload in laboratory research. Supplied as a powder in small mass quantities, it is intended for in vitro and conjugation studies.
  • Cytotoxic maytansinoid metabolite and tubulin inhibitor.
  • Suitable as an ADC payload or for conjugation studies.
  • Supplied as a powder in small mass quantities for research use.
  • CAS number 796073-68-2 for unambiguous identification.
  • Reported molecular weight 826.46 g/mol and formula C39H56ClN3O10S2.