Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Leica Microsystems DM500 Educational Microscope

Catalog No. 01672685 Shop All Leica Microsystems Products
Change view
Click to view available options
Head Style:
Binocular, 45° Inclined
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Head Style
01-672-685 Binocular, 45° Inclined
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-672-685 Supplier Leica Microsystems Supplier No. 8122671
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Leica DM500 microscope is a user-friendly educational tool, perfect for classrooms. It provides clear imaging, fostering a deeper understanding of the microscopic world for students. Ideal for educational institutions seeking a reliable and versatile microscope.

The Leica DM500 microscope is an excellent educational tool, combining precision optics with user-friendly design. Tailored for educational settings, this microscope offers clear and detailed imaging for students to explore the world of microscopy with ease. Its intuitive features make it perfect for classroom use, allowing students to engage in hands-on learning experiences. The Leica DM500 is an ideal choice for educational institutions seeking a reliable and versatile microscope to enhance the learning journey and foster a deeper understanding of the microscopic world.

  • DM 500 Right Hand Stage Stand with 4 position nosepiece
  • Universal Power Supply
  • USB Power with integrated connector
  • Mechanical stage with non-extending rack and slide holder
  • Pre-Centered and Pre-Focused Abbe condenser with slot for Phase and Darkfield Sliders
  • Abbe Condenser has Magnification labels for proper aperture setting magnifications
  • LED illumination with continuous intensity control
  • Built-in handle and cord wrap
  • 45-degree Binocular EZ Tube with integrated 10X/20 eyepieces and Pre-Set Diopters
  • Plan Objective 4X/0.10NA, 26.2mm W.D.
  • Plan Objective 10X/0.22 NA, 7.8mm W.D.
  • Plan Objective 40X/0.65 NA, 0.31MM W.D.
  • Plan Objective 100x/1.25 NA, 0.10MM W.D., Oil Immersion, 10ml oil
  • Dust cover
Includes:
  • 13613207 DM500 Brtfld 4 obj Standard Plan outfit
  • 13613900 U.S. power cord
TRUSTED_SUSTAINABILITY

Specifications

Eyepiece 10X/20 mm, with pointer
Head Style Binocular, 45° Inclined
Lighting Type LED, Transmitted Light
Magnification Power 4X, 10X, 40X, 100X
Model DM500
Objective Types Plan Achromatic
Product Type Educational Microscope
Voltage 100/240 V
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.