Learn More
New England Biolabs, Inc. 5-methyl-dCTP – 1 µmol
SDP
Catalog No. 504410
Encompass
Cytosine modification at carbon 5 (C5) represents an important epigenetic modification. It is also believed to be the starting substrate for the Ten-Eleven Translocation (TET) family of enzymes and their associated oxidation pathways.
Catalog No. 50-441-0 Supplier New England Biolabs, Inc. Supplier No. N0356S
Cytosine modification at carbon 5 (C5) represents an important epigenetic modification. It is also believed to be the starting substrate for the Ten-Eleven Translocation (TET) family of enzymes and their associated oxidation pathways.