Learn More
Leica Biosystems Leica DM6 B Upright Fluorescence Microscope - Motorized Research Imaging System
Supplier:  Leica Biosystems 11889186
Encompass_Preferred
The Leica DM6 B is a motorized upright fluorescence microscope engineered for advanced life-science and pathology imaging. It combines automation, synchronized component control, and workflow-driven software to streamline acquisition, enable large-area mosaic imaging, and support specialized configurations such as laser microdissection and fixed-stage electrophysiology.
  • Optimized for tissue, plant, and pathology imaging.
  • Supports fluorescence and brightfield imaging modalities.
  • Workflow-driven automation reduces manual tasks and increases throughput.
  • Controller support for fast mosaic imaging and synchronized component control.
  • Configurable for laser microdissection, computational clearing, and vibration-free fixed-stage electrophysiology.
  • Compatible with advanced navigator and image analysis software to support high-quality data and reproducible workflows.
Catalog No. NC3365962
Add to Cart
Request a Quote