Learn More
Leica Microsystems DM750 Educational Microscope with Eyepiece Pointers and Koehler Illumination PROMO
Catalog No. 01672680PM Shop All Leica Microsystems Products
Change view
Click to view available options
Magnification Power:
4X, 10X, 40X, 100X
Catalog No. Magnification Power
01-672-680PM 4X, 10X, 40X, 100X
1 options
Catalog No. 01-672-680PM Supplier Leica Microsystems Supplier No. 81226762026PROMO
Get up to 15% Off Leica Microscopes
View all promotions
Add to Cart
Add to Cart
The Leica DM750 microscope offers educational excellence with its versatile design. It includes an interactive eyepiece pointer, Koehler illumination, and precision optics for clear observations. Elevate your classroom experience with this powerful and user-friendly microscope.
The Leica DM750 microscope is a versatile and powerful tool designed for educational excellence. Tailored for optimal learning experiences, it features an eyepiece pointer for interactive teaching and Koehler illumination for enhanced sample visibility. With precision optics and user-friendly design, the Leica DM750 ensures clear and detailed observations, making it an ideal choice for educators and students alike. Elevate your classroom experience with this microscope that combines educational functionality and advanced features for a comprehensive and engaging exploration of the microscopic world.
  • DM 750 Right Hand Stage Stand with Standard Illumination
  • Add on Lens for Condenser
  • 4 Position nosepiece
  • Universal Power Supply plus USB Power with integrated connector to power a camera
  • Mechanical stage with non extending rack and slide holder
  • LED illumination
  • Auto Off
  • Built in handle and cord wrap
  • Dust Cover
  • User Manual
  • 45 degree Binocular EZ Tube with integrated 10X/20 eyepieces and Pre Set Diopters with Pointer on the right side
  • Abbe Condenser 0.9Dry/1.25 Oil with slot for Phase and Darkfield Sliders and color coded for matching Objectives
  • Plan 4X/0.10NA, 26.2mmW.D.
  • Plan 10X/0.22 NA, 7.8mm W.D.
  • Plan 40X/0.65 NA, 0.31MM W.D.
  • Plan 100x/1.25 NA, 0.10MM W.D.
  • Immersion Oil
Includes:
  • 13613406 DM750 BF 4Obj Plan Std EZ t Outf., pntr
  • 13613900 U.S. power cord
TRUSTED_SUSTAINABILITY
Specifications
Eyepiece 10X/20 mm
Head Style Binocular, 45° Inclined
Lighting Type LED, Transmitted Light
Magnification Power 4X, 10X, 40X, 100X
Model DM750
Objective Types Plan Achromatic
Product Type Educational Microscope
Voltage 100/240 V