Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Leica Microsystems DM750 Educational Microscope with Eyepiece Pointers and Koehler Illumination

Catalog No. 01672680 Shop All Leica Microsystems Products
Change view
Click to view available options
Head Style:
Binocular, 45° Inclined
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Head Style
01-672-680 Binocular, 45° Inclined
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-672-680 Supplier Leica Microsystems Supplier No. 8122676
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Leica DM750 microscope offers educational excellence with its versatile design. It includes an interactive eyepiece pointer, Koehler illumination, and precision optics for clear observations. Elevate your classroom experience with this powerful and user-friendly microscope.

The Leica DM750 microscope is a versatile and powerful tool designed for educational excellence. Tailored for optimal learning experiences, it features an eyepiece pointer for interactive teaching and Koehler illumination for enhanced sample visibility. With precision optics and user-friendly design, the Leica DM750 ensures clear and detailed observations, making it an ideal choice for educators and students alike. Elevate your classroom experience with this microscope that combines educational functionality and advanced features for a comprehensive and engaging exploration of the microscopic world.

  • DM 750 Right Hand Stage Stand with Standard Illumination
  • Add on Lens for Condenser
  • 4 Position nosepiece
  • Universal Power Supply plus USB Power with integrated connector to power a camera
  • Mechanical stage with non extending rack and slide holder
  • LED illumination
  • Auto Off
  • Built in handle and cord wrap
  • Dust Cover
  • User Manual
  • 45 degree Binocular EZ Tube with integrated 10X/20 eyepieces and Pre Set Diopters with Pointer on the right side
  • Abbe Condenser 0.9Dry/1.25 Oil with slot for Phase and Darkfield Sliders and color coded for matching Objectives
  • Plan 4X/0.10NA, 26.2mmW.D.
  • Plan 10X/0.22 NA, 7.8mm W.D.
  • Plan 40X/0.65 NA, 0.31MM W.D.
  • Plan 100x/1.25 NA, 0.10MM W.D.
  • Immersion Oil
Includes:
  • 13613406 DM750 BF 4Obj Plan Std EZ t Outf., pntr
  • 13613900 U.S. power cord
TRUSTED_SUSTAINABILITY

Specifications

Eyepiece 10X/20 mm
Head Style Binocular, 45° Inclined
Lighting Type LED, Transmitted Light
Magnification Power 4X, 10X, 40X, 100X
Model DM750
Objective Types Plan Achromatic
Product Type Educational Microscope
Voltage 100/240 V
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.