Learn More
Leica Microsystems DM750 Educational Microscope with Koehler Illumination PROMO
Catalog No. 01672682PM Shop All Leica Microsystems Products
Change view
Click to view available options
Magnification Power:
4X, 10X, 40X, 100X
Catalog No. Magnification Power
01-672-682PM 4X, 10X, 40X, 100X
1 options
Catalog No. 01-672-682PM Supplier Leica Microsystems Supplier No. 81226742026PROMO
Get up to 15% Off Leica Microscopes
View all promotions
Add to Cart
Add to Cart
The Leica DM750 microscope is a top educational tool, offering precision optics, advanced features, and Koehler illumination for clear observations. Durable, user-friendly, and technologically advanced, it elevates the classroom experience.
The Leica DM750 microscope is an outstanding educational tool, combining precision optics with advanced features tailored for learning environments. With Koehler illumination ensuring optimal sample visibility, this microscope provides clear and detailed observations, enhancing the educational experience. Designed for ease of use and durability, the Leica DM750 is an ideal choice for educators seeking a reliable and technologically advanced microscope to inspire and engage students in the fascinating world of microscopy. Elevate your classroom experience with the clarity and educational functionality of the Leica DM750.
  • DM 750 Right Hand Stage Stand with Standard Illumination
  • Add on Lens for Condenser
  • 4 Position nosepiece
  • Universal Power Supply plus USB Power with integrated connector
  • Mechanical stage with non extending rack and slide holder
  • LED illumination
  • Auto Off
  • Built in handle and cord wrap
  • Dust Cover
  • User Manual
  • 45 degree Binocular EZ Tube with integrated 10X/20 eyepieces and preset diopters
  • Abbe Condenser 0.9Dry/1.25 Oil with slot for Phase and Darkfield Sliders and colour-coded labels for matching Objective magnifications
  • Plan 4X/0.10NA, 26.2mmW.D.
  • Plan 10X/0.22 NA, 7.8mm W.D.
  • Plan 40X/0.65 NA, 0.31MM W.D.
  • Plan 100x/1.25 NA, 0.10MM W.D.
  • Oil Immersion Oil
Includes:
  • 13613403 DM 750 BF 4 Obj Plan Std EZ Tube Outfit
  • 13613900 U.S. power cord
TRUSTED_SUSTAINABILITY
Specifications
Eyepiece 10X/20 mm
Head Style Binocular, 45° Inclined
Lighting Type LED, Transmitted Light
Magnification Power 4X, 10X, 40X, 100X
Model DM750
Objective Types Plan
Product Type Educational Microscope
Voltage 100/240 V