Learn More
Medchemexpress LLC Dma-135 hydrochloride | 2237925-62-9 | 98.0% | C16H18ClN7O | 100 MG
SDP
Catalog No. 5000355995
Encompass_Preferred
DMA-135 hydrochloride inhibits enterovirus 71 (EV71) IRES-dependent translation and replication. It binds to enterovirus 71 (EV71) SLII domain with moderately high affinity (KD= 520 nM) and has no significant toxicity in cell-based studies.
  • Inhibits enterovirus 71 (EV71) IRES-dependent translation and replication
  • Binds to EV71 SLII domain with moderate affinity (KD= 520 nM)
  • No significant toxicity in cell-based studies
  • Induces conformational changes in RNA structure
  • Stabilizes DMA135-RNA-AUF1 ternary complex
  • Inhibits EV71 with an IC50 of 7.54 μM
Catalog No. 50-003-55995 Supplier Medchemexpress LLC Supplier No. HY145932100MG
DMA-135 hydrochloride inhibits enterovirus 71 (EV71) IRES-dependent translation and replication. It binds to enterovirus 71 (EV71) SLII domain with moderately high affinity (KD= 520 nM) and has no significant toxicity in cell-based studies.
  • Inhibits enterovirus 71 (EV71) IRES-dependent translation and replication
  • Binds to EV71 SLII domain with moderate affinity (KD= 520 nM)
  • No significant toxicity in cell-based studies
  • Induces conformational changes in RNA structure
  • Stabilizes DMA135-RNA-AUF1 ternary complex
  • Inhibits EV71 with an IC50 of 7.54 μM