Learn More
Medchemexpress LLC DMA-135 hydrochloride | 2237925-62-9 | 98.0% | C16H18ClN7O | 50 MG
SDP
Catalog No. 5000356297
Encompass_Preferred
DMA-135 hydrochloride is an inhibitor targeting enterovirus 71 (EV71) IRES-dependent translation and replication. It binds to the EV71 SLII domain with moderately high affinity (KD= 520 nM) and shows no significant toxicity in cell-based studies. It induces conformational changes in the RNA structure and stabilizes the ternary complex DMA135-RNA-AUF1, inhibiting the translation and replication of EV71 with an IC50 of 7.54 μM.
  • Targets enterovirus 71 (EV71) IRES-dependent translation and replication.
  • Binds to the EV71 SLII domain with moderately high affinity.
  • Induces conformational changes in RNA structure.
  • Stabilizes the ternary complex DMA135-RNA-AUF1.
  • Inhibits translation and replication of EV71 with an IC50 of 7.54 μM.
  • Demonstrates no significant toxicity in cell-based studies.
Catalog No. 50-003-56297 Supplier Medchemexpress LLC Supplier No. HY14593250MG
DMA-135 hydrochloride is an inhibitor targeting enterovirus 71 (EV71) IRES-dependent translation and replication. It binds to the EV71 SLII domain with moderately high affinity (KD= 520 nM) and shows no significant toxicity in cell-based studies. It induces conformational changes in the RNA structure and stabilizes the ternary complex DMA135-RNA-AUF1, inhibiting the translation and replication of EV71 with an IC50 of 7.54 μM.
  • Targets enterovirus 71 (EV71) IRES-dependent translation and replication.
  • Binds to the EV71 SLII domain with moderately high affinity.
  • Induces conformational changes in RNA structure.
  • Stabilizes the ternary complex DMA135-RNA-AUF1.
  • Inhibits translation and replication of EV71 with an IC50 of 7.54 μM.
  • Demonstrates no significant toxicity in cell-based studies.