Learn More
Anton Paar DMA 4002, 5002, and 6002 modular benchtop density meters comply with ASTM D4052, D5002, D1250, ISO 12185, and pharmacopeias. They ensure data integrity and traceability, supporting regulations such as 21 CFR Part 11.
Catalog No. 5000580480
Encompass_Preferred
SPECIFICATIONSMeasuring range:Density: 0 g/cm³ to 3 g/cm³Temperature: 0°C to 100 °C (32 °F to 212 °F)*Accuracy:Density: 0.000005 g/cm³ Temperature: 0.01 °C (0.02 °F) Repeatability, s.d.Density: 0.000001 g/cm³Temperature: 0.001 °C (0.002 °F)Reproducibility, s.d.Density: 0.0000005 g/cm³Digital ResolutionDensity: 0.000001 g/cm³
Catalog No. 50-005-80480 Supplier Anton Paar Supplier No. 325201
Please contact your sales representative for a quote or further ordering instructions.
SPECIFICATIONSMeasuring range:Density: 0 g/cm³ to 3 g/cm³Temperature: 0°C to 100 °C (32 °F to 212 °F)*Accuracy:Density: 0.000005 g/cm³ Temperature: 0.01 °C (0.02 °F) Repeatability, s.d.Density: 0.000001 g/cm³Temperature: 0.001 °C (0.002 °F)Reproducibility, s.d.Density: 0.0000005 g/cm³Digital ResolutionDensity: 0.000001 g/cm³