Learn More
Anton Paar DMA 4002, 5002, and 6002 modular benchtop density meters comply with ASTM D4052, D5002, D1250, ISO 12185, and pharmacopeias. They ensure data integrity and traceability, supporting regulations such as 21 CFR Part 11.
Catalog No. 5000580539
Encompass_Preferred
SPECIFICATIONSMeasuring range:Density: 0 g/cm³ to 3 g/cm³Temperature: 0°C to 100 °C (32 °F to 212 °F)*Accuracy:Density: 0.00001 g/cm³ (0 g/cm³ to 1.05 g/cm³, 15°C to 20°C) 0.00005 g/cm³ (Full Range)Temperature: 0.01 °C (0.02 °F) ( 15°C to 20°C)0.015 °C (0.03 °F) (Full Range) Repeatability, s.d.Density: 0.000003 g/cm³Temperature: 0.005 °C (0.01 °F)Reproducibility, s.d.Density: 0.000005 g/cm³Digital ResolutionDensity: 0.000005 g/cm³
Catalog No. 50-005-80539 Supplier Anton Paar Supplier No. 325159
Please contact your sales representative for a quote or further ordering instructions.
SPECIFICATIONSMeasuring range:Density: 0 g/cm³ to 3 g/cm³Temperature: 0°C to 100 °C (32 °F to 212 °F)*Accuracy:Density: 0.00001 g/cm³ (0 g/cm³ to 1.05 g/cm³, 15°C to 20°C) 0.00005 g/cm³ (Full Range)Temperature: 0.01 °C (0.02 °F) ( 15°C to 20°C)0.015 °C (0.03 °F) (Full Range) Repeatability, s.d.Density: 0.000003 g/cm³Temperature: 0.005 °C (0.01 °F)Reproducibility, s.d.Density: 0.000005 g/cm³Digital ResolutionDensity: 0.000005 g/cm³