Learn More
Medchemexpress LLC HY-15621 5mg Medchemexpress, DMA CAS:188860-26-6 Purity:>98%
SDP
Catalog No. 502081752
Encompass_Preferred
Medchemexpress, HY-15621 5mg DMA CAS:188860-26-6 DMA is a fluorescent compound (λex=340 nm, λem=478 nm). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1752 Supplier Medchemexpress LLC Supplier No. HY156215MG
Medchemexpress, HY-15621 5mg DMA CAS:188860-26-6 DMA is a fluorescent compound (λex=340 nm, λem=478 nm). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.