Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences DMA N, N-Dimethylaminoethyl Methacrylate 450 ML
SDP

Catalog No. 50980365
Encompass_Preferred

CAS #2867-47-2 C₈H₁₅NO₂ F.W. 157.21
Boiling point: 76°C

A water soluble monomer.

Catalog No. 50-980-365 Supplier Electron Microscopy Sciences Supplier No. 13350
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS #2867-47-2 C₈H₁₅NO₂ F.W. 157.21
Boiling point: 76°C

A water soluble monomer.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.