Learn More
Medchemexpress LLC HY-15621A 5mg Medchemexpress, DMA (trihydrochloride) CAS:2095832-33-8 Purity:>98%
SDP
Catalog No. 502081753
Encompass_Preferred
Medchemexpress, HY-15621A 5mg DMA (trihydrochloride) CAS:2095832-33-8 DMA trihydrochloride is a fluorescent compound (λex=340 nm, λem=478 nm). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1753 Supplier Medchemexpress LLC Supplier No. HY15621A5MG
Medchemexpress, HY-15621A 5mg DMA (trihydrochloride) CAS:2095832-33-8 DMA trihydrochloride is a fluorescent compound (λex=340 nm, λem=478 nm). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.