Learn More
Apexbio Technology LLC DMAB-anabaseine dihydrochloride 154149-38-9 10mg
SDP
Supplier:  Apexbio Technology LLC B699010
Encompass_Preferred
DMAB-anabaseine dihydrochloride (CAS 154149-38-9) is a small-molecule partial agonist and antagonist targeting neuronal 7 nicotinic acetylcholine receptors (nAChRs) It is designed to modulate receptor-mediated neurotransmission thereby influencing cognitive-related neuronal signaling pathways DMAB-anabaseine dihydrochloride exerts its biological activity primarily through selective partial activation of 7 nAChRs and antagonism at other subtypes such as 4 2 Based on these pharmacological properties DMAB-anabaseine dihydrochloride holds research potential in studies of nicotinic receptor function neural signaling mechanisms and cognitive-related conditions
Catalog No. 50-000-90241