Learn More
Cayman Chemical DMABA-d10 NHS ester, Purity: ≥99% deuterated forms (d1-d10), CAS: 1175002-05-7, 1 mg
SDP
Catalog No. 502951340
Encompass_Preferred
DMABA-d10 NHS ester is intended for use in combination with DMABA NHS ester (Item No. 11216) as a derivatizing reagent for PE lipids in order to facilitate MS characterization and to quantify relative changes in their abundance
Catalog No. 50-295-1340 Supplier Cayman Chemical Supplier No. 112191 MG
DMABA-d10 NHS ester is intended for use in combination with DMABA NHS ester (Item No. 11216) as a derivatizing reagent for PE lipids in order to facilitate MS characterization and to quantify relative changes in their abundance