Learn More
Cayman Chemical DMABA-d4 NHS ester, Purity: ≥99% deuterated forms (d1-d4), CAS: 1175002-03-5, 5 mg
SDP
Catalog No. 502954012
Encompass_Preferred
DMABA-d4 NHS ester is intended for use in combination with DMABA NHS ester (Item No. 11216) as a derivatizing reagent for PE lipids in order to facilitate MS characterization and to quantify relative changes in their abundance
Catalog No. 50-295-4012 Supplier Cayman Chemical Supplier No. 112175 MG
DMABA-d4 NHS ester is intended for use in combination with DMABA NHS ester (Item No. 11216) as a derivatizing reagent for PE lipids in order to facilitate MS characterization and to quantify relative changes in their abundance