Learn More
Cayman Chemical DMABA-d6 NHS ester, Purity: ≥99% deuterated form (d1-d6), CAS: 1175002-04-6, 10 mg
SDP
Catalog No. 502951622
Encompass_Preferred
DMABA-d6 NHS ester is intended for use in combination with DMABA NHS ester (Item No. 11216) as a derivatizing reagent for PE lipids in order to facilitate MS characterization and to quantify relative changes in their abundance
Catalog No. 50-295-1622 Supplier Cayman Chemical Supplier No. 1121810 MG
DMABA-d6 NHS ester is intended for use in combination with DMABA NHS ester (Item No. 11216) as a derivatizing reagent for PE lipids in order to facilitate MS characterization and to quantify relative changes in their abundance