Learn More
Cayman Chemical DMABA NHS ester, Purity: ≥98%, CAS: 58068-85-2, 100 mg
SDP
Catalog No. 502952529
Encompass_Preferred
A reagent that reacts with the primary amine group of PE lipids; facilitates the use of electrospray tandem MS for the detection of diacyl, ether, and plasmalogen PE lipid
Catalog No. 50-295-2529 Supplier Cayman Chemical Supplier No. 11216100 MG
A reagent that reacts with the primary amine group of PE lipids; facilitates the use of electrospray tandem MS for the detection of diacyl, ether, and plasmalogen PE lipid