Learn More
Medchemexpress LLC DMABA NHS ester | 58068-85-2 | MFCD16875522 | 99.0% | 262.26 g/mol | C13H14N2O4 | 25 MG
SDP
Catalog No. 5000308295
Encompass_Preferred
DMABA NHS ester is the N-hydroxysuccinimide ester of DMABA, a reactive labeling reagent used to derivatize primary amines for detection and analysis in biochemical workflows. It is supplied as a high-purity solid and is suitable for research applications requiring amine-specific tagging.
  • Reactive toward primary amines for labeling and conjugation.
  • Suitable for phospholipid, peptide, and protein derivatization for MS or fluorescence detection.
  • High purity ensures minimal side products during reactions.
  • Supplied as a solid for convenient storage and handling.
  • Store sealed, away from moisture; long-term storage at -20°C recommended.
Catalog No. 50-003-08295 Supplier Medchemexpress LLC Supplier No. HY11843125MG
DMABA NHS ester is the N-hydroxysuccinimide ester of DMABA, a reactive labeling reagent used to derivatize primary amines for detection and analysis in biochemical workflows. It is supplied as a high-purity solid and is suitable for research applications requiring amine-specific tagging.
  • Reactive toward primary amines for labeling and conjugation.
  • Suitable for phospholipid, peptide, and protein derivatization for MS or fluorescence detection.
  • High purity ensures minimal side products during reactions.
  • Supplied as a solid for convenient storage and handling.
  • Store sealed, away from moisture; long-term storage at -20°C recommended.