Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences DMAE (2-Dimethylaminoethanol) 25 ML
SDP

Catalog No. 50980364
Encompass_Preferred

CAS #108-01-0 C₄H₁₁NO F.W. 89.14
Boiling point: 132-134°C

A curing agent for epoxy resins.

Catalog No. 50-980-364 Supplier Electron Microscopy Sciences Supplier No. 13300
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS #108-01-0 C₄H₁₁NO F.W. 89.14
Boiling point: 132-134°C

A curing agent for epoxy resins.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.