Learn More
Cayman Chemical DMA (hydrochloride), Purity: ≥98%, CAS: 2095832-33-8, 10 mg
SDP
Catalog No. 502988872
Encompass_Preferred
A radi; oprotective agent; decreases radiation-induced lethality without affecting radiation-induced tumor regression in an Ehrlich murine spontaneous adenocarcinoma model at 300 mg/kg; prevents radiation-induced damage of intestinal, hepatic, and splenic tissues, increases in splenic MDA production, and decreases in splenic SOD activity in a B16/F10 murine melanoma model at 50 mg/kg
Catalog No. 50-298-8872 Supplier Cayman Chemical Supplier No. 3041610 MG
A radi; oprotective agent; decreases radiation-induced lethality without affecting radiation-induced tumor regression in an Ehrlich murine spontaneous adenocarcinoma model at 300 mg/kg; prevents radiation-induced damage of intestinal, hepatic, and splenic tissues, increases in splenic MDA production, and decreases in splenic SOD activity in a B16/F10 murine melanoma model at 50 mg/kg