Learn More
Apexbio Technology LLC DMAP 1122-58-3 100g
SDP
Catalog No. 5000097142
Encompass_Preferred
4-Dimethylaminopyridine (DMAP) (CAS 1122-58-3) is a pyridine-based aromatic amine commonly utilized as a nucleophilic catalyst DMAP exerts its catalytic activity through nucleophilic activation transiently coordinating to acyl intermediates to facilitate efficient acyl transfer reactions Based on these properties DMAP holds research potential in the synthetic modification of peptides nucleotides and bioactive molecules drug synthesis derivatization of natural compounds and the preparation of bioactive molecular scaffolds for therapeutic development
Catalog No. 50-000-97142 Supplier Apexbio Technology LLC Supplier No. A7055100000
Add to Cart
Add to Cart
4-Dimethylaminopyridine (DMAP) (CAS 1122-58-3) is a pyridine-based aromatic amine commonly utilized as a nucleophilic catalyst DMAP exerts its catalytic activity through nucleophilic activation transiently coordinating to acyl intermediates to facilitate efficient acyl transfer reactions Based on these properties DMAP holds research potential in the synthetic modification of peptides nucleotides and bioactive molecules drug synthesis derivatization of natural compounds and the preparation of bioactive molecular scaffolds for therapeutic development