Learn More
Apexbio Technology LLC DMAP 1122-58-3 25g
SDP
Catalog No. 502240297
Encompass_Preferred
4-Dimethylaminopyridine (DMAP) (CAS 1122-58-3) is a pyridine-based aromatic amine commonly utilized as a nucleophilic catalyst DMAP exerts its catalytic activity through nucleophilic activation transiently coordinating to acyl intermediates to facilitate efficient acyl transfer reactions Based on these properties DMAP holds research potential in the synthetic modification of peptides nucleotides and bioactive molecules drug synthesis derivatization of natural compounds and the preparation of bioactive molecular scaffolds for therapeutic development
This item is not returnable. View return policy
Catalog No. 50-224-0297 Supplier Apexbio Technology LLC Supplier No. A705525000
4-Dimethylaminopyridine (DMAP) (CAS 1122-58-3) is a pyridine-based aromatic amine commonly utilized as a nucleophilic catalyst DMAP exerts its catalytic activity through nucleophilic activation transiently coordinating to acyl intermediates to facilitate efficient acyl transfer reactions Based on these properties DMAP holds research potential in the synthetic modification of peptides nucleotides and bioactive molecules drug synthesis derivatization of natural compounds and the preparation of bioactive molecular scaffolds for therapeutic development