Learn More
TARGETMOL CHEMICALS INC DMAPP triammonium 1MG
SDP
Catalog No. 502876114
Encompass_Preferred
DMAPP triammonium (Dimethylallyl diphosphate triammonium) is one of the prerequisite substances for an isoprenoid involved in the mevalonate (MVA) pathway, which can be used in the study of metabolism-related diseases. Purity 95.38%
Catalog No. 50-287-6114 Supplier TARGETMOL CHEMICALS INC Supplier No. T385701MG
DMAPP triammonium (Dimethylallyl diphosphate triammonium) is one of the prerequisite substances for an isoprenoid involved in the mevalonate (MVA) pathway, which can be used in the study of metabolism-related diseases. Purity 95.38%