Learn More
Medchemexpress LLC HY-16172 5mg Medchemexpress, DMAPT CAS:870677-05-7 Purity:>98%
SDP
Catalog No. 501969968
Encompass_Preferred
Medchemexpress, HY-16172 5mg DMAPT CAS:870677-05-7 DMAPT (Dimethylamino Parthenolide), a water soluble analogue of Parthenolide (PTL), is an oral active NF-κB inhibitor, with a LD50 of 1.7 μM for cell population in AML cells. Has potential anti-cancer and anti-metastatic effect. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9968 Supplier Medchemexpress LLC Supplier No. HY161725MG
Medchemexpress, HY-16172 5mg DMAPT CAS:870677-05-7 DMAPT (Dimethylamino Parthenolide), a water soluble analogue of Parthenolide (PTL), is an oral active NF-κB inhibitor, with a LD50 of 1.7 μM for cell population in AML cells. Has potential anti-cancer and anti-metastatic effect. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.