Learn More
Abcam DMAPT, NF-kappaB inhibitor, 5MG
Catalog No. 502704863
Encompass_Preferred
MW 293.41 g/mol. NF-κB inhibitor. Reduces NF-κB DNA binding. Shows antiproliferative effects (IC₅₀ = 5-10 μM). Parthenolide (ab120849) analog. Epigenetic modulator, functions NFκB-dependently and independently. Generates ROS and shows antitumor effects in vivo. Orally active.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4863 Supplier Abcam Supplier No. AB1461895MG
MW 293.41 g/mol. NF-κB inhibitor. Reduces NF-κB DNA binding. Shows antiproliferative effects (IC₅₀ = 5-10 μM). Parthenolide (ab120849) analog. Epigenetic modulator, functions NFκB-dependently and independently. Generates ROS and shows antitumor effects in vivo. Orally active.
The product is subject to the following: Abcam Restricted Use Statement