Learn More
Apexbio Technology LLC Demethylasterriquinone B1(Synonyms: Demethylasterriquinone B-1, DMAQ-B1, Demethylasterriquinone-B1, DMAQ B1, Demethylasterriquinone derivative B1), 5mg, CAS: 78860-34-1.
SDP
Catalog No. 5000060192
Encompass_Preferred
Demethylasterriquinone B1 (CAS 78860-34-1) is a selective insulin receptor (IR) agonist that acts by promoting tyrosine phosphorylation within the insulin receptor -subunit This phosphorylation event subsequently activates downstream signaling pathways particularly phosphatidylinositol 3-kinase (PI3K) and AKT Demethylasterriquinone B1 has utility as a research tool for investigating insulin signaling cascades glucose metabolism and related metabolic regulatory mechanisms thus facilitating studies aimed at understanding insulin sensitivity and metabolic diseases
Catalog No. 50-000-60192 Supplier Apexbio Technology LLC Supplier No. B68695
Demethylasterriquinone B1 (CAS 78860-34-1) is a selective insulin receptor (IR) agonist that acts by promoting tyrosine phosphorylation within the insulin receptor -subunit This phosphorylation event subsequently activates downstream signaling pathways particularly phosphatidylinositol 3-kinase (PI3K) and AKT Demethylasterriquinone B1 has utility as a research tool for investigating insulin signaling cascades glucose metabolism and related metabolic regulatory mechanisms thus facilitating studies aimed at understanding insulin sensitivity and metabolic diseases