Learn More
FUJIFILM BIOSCIENCES INC DME High Glucose Liquid
Supplier:  FUJIFILM BIOSCIENCES INC 9031500ML
Encompass
Dulbecco’s Modified Eagle’s Medium (DMEM) is a modification of Basal Medium Eagle (BME) containing two to fourfold higher concentrations of glucose, amino acids, and vitamins to better support highly proliferative cells such as embryonic cells.
Catalog No. NC0003787