Learn More
FUJIFILM Biosciences DME High Glucose Liquid
Catalog No. 5000450299
Encompass
Dulbecco s Modified Eagle s Medium DMEM is a modification of Basal Medium Eagle BME containing two to fourfold higher concentrations of glucose amino acids and vitamins to better support highly proliferative cells such as embryonic cells
Catalog No. 50-004-50299 Supplier FUJIFILM Biosciences Supplier No. 90311L
Dulbecco s Modified Eagle s Medium DMEM is a modification of Basal Medium Eagle BME containing two to fourfold higher concentrations of glucose amino acids and vitamins to better support highly proliferative cells such as embryonic cells