Learn More
FUJIFILM BIOSCIENCES INC DME High Glucose w/oLGlutamin
Supplier:  FUJIFILM BIOSCIENCES INC 90241L
Encompass
Dulbeccos Modified Eagles medium DME is a modification of Basal Medium Eagle BME and contains a fourfold increase of amino acids vitamins and nutrients to provide support for a wide variety of cellLines
Catalog No. 50-004-50260