Learn More
FUJIFILM Biosciences DME High Glucose w/oLGlutamine
Catalog No. 5000450193
Encompass
Dulbeccos Modified Eagles medium DME is a modification of Basal Medium Eagle BME and contains a fourfold increase of amino acids vitamins and nutrients to provide support for a wide variety of cellLines
Catalog No. 50-004-50193 Supplier FUJIFILM Biosciences Supplier No. 9024500ML
Dulbeccos Modified Eagles medium DME is a modification of Basal Medium Eagle BME and contains a fourfold increase of amino acids vitamins and nutrients to provide support for a wide variety of cellLines