Learn More
Quality Biological Inc DMEM with 1.5 g/L Sodium Bicarbonate and L-Glutamine, 500 mL
SDP
Catalog No. 502209746
Encompass_Preferred
DMEM (Dulbeccos Modified Eagle Medium) is a widely used basal medium for supporting the growth of many different mammalian cells. Cells successfully cultured in DMEM include primary fibroblasts neurons glial cells HUVECs and smooth muscle cells. We offer a variety of DMEM modifications for a range of cell culture applications.
With High Glucose
With Sodium Bicarbonate
Without HEPES
Manufactured under an ISO 9001:2015 certified quality management system.
Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.
Catalog No. 50-220-9746 Supplier Quality Biological Inc Supplier No. 112319101
DMEM (Dulbeccos Modified Eagle Medium) is a widely used basal medium for supporting the growth of many different mammalian cells. Cells successfully cultured in DMEM include primary fibroblasts neurons glial cells HUVECs and smooth muscle cells. We offer a variety of DMEM modifications for a range of cell culture applications.
With High Glucose
With Sodium Bicarbonate
Without HEPES
Manufactured under an ISO 9001:2015 certified quality management system.
Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.