Learn More
Sigma Aldrich Fine Chemicals Biosciences DMEM Complete Medium, with 2mM L-Glut, Sodium Pyruvate, and 10% FBS DMEM Complete Medium contains DMEM high glucose medium with 2mM L-glut, sodium pyruvate, and 10% U.S. origin FBS. DMEM
Catalog No. NC2125513
Encompass
DMEM Complete Medium, with 2mM L-Glut, Sodium Pyruvate, and 10% FBS DMEM Complete Medium contains DMEM high glucose medium with 2mM L-glut, sodium pyruvate, and 10% U.S. origin FBS. DMEM Complete Medium is a convenient sterile solution for the culture of primary mammalian cells including mouse and chicken cells, and a variety of normal and transformed cells. |
This item is not returnable. View return policy
Catalog No. NC2125513 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. SLM241B
DMEM Complete Medium, with 2mM L-Glut, Sodium Pyruvate, and 10% FBS DMEM Complete Medium contains DMEM high glucose medium with 2mM L-glut, sodium pyruvate, and 10% U.S. origin FBS. DMEM Complete Medium is a convenient sterile solution for the culture of primary mammalian cells including mouse and chicken cells, and a variety of normal and transformed cells. |