Learn More
ATHENA ES DMEM/F12 1:1, 500 ml
Catalog No. NC3870163
Encompass_Preferred
DMEM/F12 is a serum-free medium formulation for general use. This formulation is a 1:1 blend of DMEM and Ham’s F12 media supplied complete and ready to use with L-glutamine, Hepes, BPE and EGF for culturing a wide range of cell types. DMEM/F12 does not contain phenol red or antibiotics.
Catalog No. NC3870163 Supplier ATHENA ES Supplier No. 0410
DMEM/F12 is a serum-free medium formulation for general use. This formulation is a 1:1 blend of DMEM and Ham’s F12 media supplied complete and ready to use with L-glutamine, Hepes, BPE and EGF for culturing a wide range of cell types. DMEM/F12 does not contain phenol red or antibiotics.