Learn More
ATHENA ES DMEM/F12, basal media
Catalog No. 5000158132
Encompass_Preferred
Dulbecco's Modified Eagle Medium: Nutrient Mixture F12 (DMEM F12) is a widely used basal medium for supporting the growth of many different mammalian cells including MDCK, glial cells, fibroblasts, human endothelial cells, and rat fibroblasts. DMEM F12 is a 1:1 mixture of DMEM and Ham's F12 media. This formulation combines DMEM's high concentrations of glucose, amino acids, and vitamins with the wide variety of components found in Ham's F12 media.
Catalog No. 50-001-58132 Supplier ATHENA ES Supplier No. 0463
Dulbecco's Modified Eagle Medium: Nutrient Mixture F12 (DMEM F12) is a widely used basal medium for supporting the growth of many different mammalian cells including MDCK, glial cells, fibroblasts, human endothelial cells, and rat fibroblasts. DMEM F12 is a 1:1 mixture of DMEM and Ham's F12 media. This formulation combines DMEM's high concentrations of glucose, amino acids, and vitamins with the wide variety of components found in Ham's F12 media.